Learn More
MyBioSource INHBA antibody
SDP
Supplier:  MyBioSource MBS82423660.2 ML
Encompass_Preferred
The MyBioSource INHBA antibody is a rabbit polyclonal antibody raised against a recombinant protein of human INHBA. This antibody detects INHBA in direct ELISAs and Western blots. INHBA is a subunit of activin and inhibin, which are involved in follicle stimulating hormone secretion, erythropoiesis, neuronal cell differentiation, and apoptosis. This antibody can be used for ELISA, WB, IHC, and neutralization. It is provided at 100 μg in PBS with 0.02% sodium azide and 50% glycerol.
Catalog No. NC2753533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.