Learn More
Cytiva MicroFunnel™ Plus Filter Funnel with Supor Membrane, 300 mL, 0.2 µm, White, Gridded, Sterile, Individually Bagged (20/pkg)
Catalog No. NC1111683 Shop All Cytiva Products
Encompass
Combined Filter Funnel and Sample Cup Revolutionizes Water System Monitoring to Eliminate the Risk of Contamination-Product performs as sample container and filter funnel all in one. -MicroFunnel Plus AP unit allows aseptic collection of the sample through the lid sample port. No need to remove lid.-MicroFunnel Plus products with Supor membrane are designed to allow sampling of hot water up to 90°C-Vented lid snaps to a liquid-tight seal and allows filtration without having to open the funnel and risk introducing contamination.-Vent filter ensures no airborne contamination is drawn into the funnel during filtration.-Attaches directly to a standard laboratory manifold or use with adapter and stopper.-Volume graduations are clearly marked for ease in measuring your sample volume.-Individually bagged and labeled for contamination control and lot traceability. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1111683 Supplier Cytiva Supplier No. 4857
Combined Filter Funnel and Sample Cup Revolutionizes Water System Monitoring to Eliminate the Risk of Contamination-Product performs as sample container and filter funnel all in one. -MicroFunnel Plus AP unit allows aseptic collection of the sample through the lid sample port. No need to remove lid.-MicroFunnel Plus products with Supor membrane are designed to allow sampling of hot water up to 90°C-Vented lid snaps to a liquid-tight seal and allows filtration without having to open the funnel and risk introducing contamination.-Vent filter ensures no airborne contamination is drawn into the funnel during filtration.-Attaches directly to a standard laboratory manifold or use with adapter and stopper.-Volume graduations are clearly marked for ease in measuring your sample volume.-Individually bagged and labeled for contamination control and lot traceability. This product is not approved for use in medical, clinical, surgical or other patient applications.