Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.35 to 19.4 mg/mL Residual Solvents Class 2 Mix A, 16 Components, (2013 Rev) In Dimethyl Sulfoxide, Restek
SDP

Catalog No. 06611926 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-611-926 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-926 Supplier Restek Corporation Supplier No. 36012
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For detecting and quantifying USP 467 class 2 residual solvents in pharmaceuticals

  • Reformulated to include cumene, added to USP 467 class 2 list in August 2013
  • Quantitatively tested to confirm composition and stability

Compounds:

  • Acetonitrile
  • Chlorobenzene
  • Cyclohexane
  • cis-1,2-Dichloroethene
  • trans-1,2-Dichloroethene
  • 1,4-Dioxane
  • Ethylbenzene
  • Isopropylbenzene (cumene)
  • Methanol
  • Methylcyclohexane
  • Methylene chloride (dichloromethane)
  • Tetrahydrofuran
  • Toulene
  • m-Xylene
  • o-Xylene
  • p-Xylene

Specifications

Solution Contains 16 components
Shipping Condition Ambient
Concentration 0.35 to 19.4 mg/mL
Certifications/Compliance Certified reference material
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.