Learn More
ProteinSimple 0.5% Methyl Cellulose Kit, 2 bottles x 100mL
Supplier: ProteinSimple 102505
0.5% Methyl Cellulose Solution, 100mL Use this concentration for the wash cycle between injections. The solution is filtered to ensure consistent viscosity to coat the capillary lumen to minimize electroosmotic flow (EOF). Conveniently pac
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.