Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized

Catalog No. 14222561 Shop All Axygen Products
Change view
Click to view available options
Closure Color:
Assorted
Blue
Clear
Green
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
14-222-561 Yellow
14-222-517 Assorted
14-222-523 Clear
14-222-532 Red
14-222-527 Green
14-222-521 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-222-561 Supplier Axygen™ Supplier No. SCT050YS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features chemical resistant O-ring

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.

  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Yellow
Closure Material Polypropylene
Capacity (Metric) 0.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Packaging 500 Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Product Type Screw Cap Tube
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.