Learn More
Invitrogen™ Mouse IgG1 Isotype Control, Alexa Fluor™ 488
Mouse Monoclonal Isotype Control
Supplier:  Invitrogen™ MG120
Encompass_Preferred
Catalog No. MG120
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG1
Monoclonal
PBS with 0.1% sodium azide
Mouse
500 μL
Whole Antibody
Not Applicable
Isotype Control
IgG1
Wet Ice
Flow Cytometry
Alexa Fluor 488
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Purified
ASR
Isotype Control (Primary)
2-8°C
Liquid
50 Tests
Alexa Fluor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.