Learn More
Leica Biosystems 0.5ml NCL-L-CD19-163
Supplier:  Leica Biosystems CD19163LCEH
Encompass
CD19Antigen BackgroundCD19 is a member of the immunoglobulin superfamily and has two Ig like domains. It is a single chain glycoprotein present on the surface of B lymphocytes and follicular dendritic cells of the hematopoietic system. CD19 is a crucial regulator in B cell development, activation and differentiation. On B cells, CD19 associates with CD21, CD81 and CD225 (Leu-13) forming a signal transduction complex. CD19 is expressed from the earliest recognizable B cell lineage stage, through development to B cell differentiation but is lost on maturation to plasma cells.DisclaimerCD19 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2330
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.