Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Adapter

Catalog No. 05414341 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
12 x 75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
05-414-341 12 x 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-414-341 Supplier Eppendorf™ Supplier No. 5825747003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • For 27 round-bottom tubes 5 mL FACS
  • For Rotor S-4-104
  • Rotor S-4x1000 round buckets and Rotor S-4x750
  • 2 pcs
TRUSTED_SUSTAINABILITY

Specifications

Type Adapter
Capacity 12 x 75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.