Learn More
Nova Biostorage 0.75ml 2D Data-Matrix Coded Screw Cap Tubes, Internal Thread V-Bottom, Precapped Light Blue Screw Caps, Micronic 96-2 Rack, High Cover, Barcoded A1-H1 Side, Gamma Irradiated, 40 Racks
SDP
Supplier:  Nova Biostorage MP52520SZ24
Encompass_Preferred
0.75ml 2D Data-Matrix coded Screw Cap tubes with Internal Thread V-bottom. Precapped with Light Blue color screw caps in Micronic 96-2 Rack and high cover. (Barcoded A1-H1 side)
Gamma Irradiated
Working volume at 21°C: 0.48ml
Quantity = 40 Racks
Catalog No. NC3861657
Only null left
Add to Cart
Request a Quote