Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Iodine Solution 0.9%, Aldon Corporation
SCI_ED

Catalog No. S28450 Shop All Aldon Products
Change view
Click to view available options
Quantity:
30 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S28450 500 mL
S28452 30 mL
S28451 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S28450 Supplier Innovating Science™ Supplier No. ADIX0132500ML
Add to Cart
Add to Cart

Approximately 0.0156M solution in H2O

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Boiling Point 100°C (aq. soln.)
Melting Point 0°C (aq. soln.)
CAS Max % 0.3959%
Physical Form Liquid or Solid
Quantity 500 mL
Flash Point Not flammable
Solubility Information Completely soluble in water
Percent Purity 0.90%
Grade Reagent
Odor Slight iodine odor
Packaging PVC Bottle
Appearance Liquid. Clear to yellow
Specific Gravity 1.0 (in water)
Vapor Pressure 14 mm in Hg (in water)
Density Vapor Density: 0.7 (Air= 1)
Chemical Name or Material Iodine Solution, 0.0156M
Show More Show Less
EINECSNumber 231-442-4
RTECSNumber NN1575000
storageNote1 Store in a cool, well-ventilated area away from incompatible substances
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.