Learn More
Description
- Achieve consistent results with built-in automatic calibration, touchless sensors and a ventilated weighing pan
- Get reliable data with the digital weighing cell and internal environment sensors
- A intuitive 7 in. glass touchscreen, digital leveling, and simplified data management is easy to use
- With an effortless installation and chemical-protective design it ensures efficiency and comfort
- Supports HID connection to PC or keyboard without drivers, and provides 2-way communication with RS232, USB, Ethernet, Bluetooth, and WiFi for seamless data transfer
Specifications
Specifications
| Capacity | 120 g |
| Display Type | Touchscreen |
| Readability | 0.01 mg |
| Type | Microbalances |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
