Learn More
Metrohm USA 01195822 ELECTRODE (SLEEVELESS
Supplier: Metrohm USA 60229020
The space-saving electrode for titration in nonaqueous media with easyClean diaphragm - with 2 m cable for automation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.