Learn More
OHAUS 6 Clip Plates
Catalog No. 01804357 Shop All OHAUS Products
Change view
Click to view available options
Holds:
12 Tubes
3 Tubes
6 Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
01-804-357 6 Tubes
01-804-385 3 Tubes
01-804-404 12 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-804-357 Supplier OHAUS Supplier No. 83041379
Add to Cart
Add to Cart
Pack of 6 metal clip plates to accommodate up to 12, 6, and 3 tubes with a diameter of 10-13 mm, 15-17 mm and 28-30 mm respectively.
  • A maximum of 8 clip plates, 4 per side.
  • It may be used with the Roto-Shake Genie, Rotator Genie, Enviro-Genie, or Incubator Genie.
TRUSTED_SUSTAINABILITY
Specifications
Holds 6 Tubes
Product Type Shaker Attachment
Diameter (Metric) 15 to 17 mm
Packaging Quantity 6 Pk.