Learn More
OHAUS Adhering Blue Mat 18 x 18 in.
Catalog No. 01804359 Shop All OHAUS Products
Click to view available options
Product Type:
Vortex Attachment
Blue adhering mat is an alternative to flask clamps or roller bars for securing vessels to shaker platform.
  • It is ideal for containers such as bottles and dishes that cannot be held with conventional clamps or roller bars.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Orbital Genie and accessories
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.