Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Multi-Position Stirrer

Catalog No. 01918309 Shop All OHAUS Products
Change view
Click to view available options
Stirring Positions:
4
9
Controller Type:
Analog
Digital
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stirring Positions Controller Type
01-918-309 4 Digital
01-918-307 4 Analog
01-918-308 9 Analog
01-918-310 9 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-918-309 Supplier OHAUS™ Supplier No. 30573799
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use the Multi-Position Stirrer for synchronized and uniform mixing.

  • Applications: Dissolution Studies, Titrations, Media/Reagent Preparations, Synchronized Stirring of multiple samples
  • Display: Independent LED displays for speed and time (on digital models); Knob with 1-10 dial markings (on non-digital models)
  • Operation: N/A
  • Communication: N/A
  • Construction: Cast aluminum housing, glass filled nylon top plate. Includes stir bars to accommodate each stirring position
  • Design Features: Speed ramping feature slowly increases motor speed for enhanced coupling and improved stirring. Microprocessor-controlled for precise speed regulation. Timer counts up or when programmed to a set time will turn off stirring and sound alarm when timer reaches zero.
TRUSTED_SUSTAINABILITY

Specifications

Speed 60 to 1400 rpm
Stirring Positions 4
Top Plate Material Glass-filled Nylon
Capacity 1 L
Display Digital LED
Shape Square
Voltage 120 V
Wattage 75 W
Height (English) 4 in.
Height (Metric) 970 mm
Length (English) 17 in.
Length (Metric) 432 mm
Top Plate Length (English) 12.5 in.
Top Plate Length (Metric) 318 mm
Top Plate Width (English) 11 in.
Top Plate Width (Metric) 279 mm
Width (English) 11 in.
Width (Metric) 279 mm
Amperage 1.3 A
Frequency 50/60 Hz
Certifications/Compliance UL, cUL
Controller Type Digital
Model STMP4DG
Temperature Range (Metric) 5°C to 40°C, 80% RH, Non-condensing
Timer 1 sec. to 160 hr.
Type Multi-Position Stirrers
Weight (English) 14 lb.
Weight (Metric) 6.3 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.