Learn More
Thermo Scientific Zeta Plus™ Encapsulated System Filter Capsule, 1020 cm2
Catalog No. 02555447
Change view
Click to view available options
Grade:
05SP01A
10SP02A
30SP02A
60SP02A
60SP03A
60SP05A
90SP05A
90SP08A
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
02-555-447 10SP02A
02-555-471 05SP01A
02-555-558 30SP02A
02-555-572 60SP02A
02-555-780 60SP03A
02-555-445 60SP05A
02-555-460 90SP05A
02-555-479 90SP08A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
The brand will transition from 3M to Thermo Scientific
Catalog No. 02-555-447 Supplier Thermo Scientific Supplier No. E1020FSA 10SP02A
Add to Cart
Add to Cart
The brand will transition from 3M to Thermo Scientific
Encapsulated System Scale‐Up Filter Capsules, EXT, with SP Series Media offer economical small‐scale clarification of pharmaceutical, biological, bioprocess and cosmetic fluids. The depth media blends filter aid, cellulose and a charged resin to adsorb negatively charged contaminants.
  • Two layers of complementary porosities provide high contaminant holding capacity, economical filtration and reliable particle reduction
  • FDA Drug Master File and USP <88> Class VI Biological Reactivity Tests eases validation and regulatory submissions
  • Self-contained, single-use capsules are available in three convenient sizes for scale-up and benchtop studies
  • Low hold-up volume for minimal fluid loss
  • Novel shape maintains the same lenticular flow path as larger capsules and cartridges
  • High contaminant holding capacity due to two layers of complementary porosities provides economical filtration and reliable particle reduction
Specifications
Disposable Single-use
Filter Type Filter Capsule
Material (Membrane) Cellulose
Sterility Non-sterile
Grade 10SP02A
Quantity 1 Ea.
Type Filter Capsule