Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Zeta Plus™ Encapsulated System Scale-Up Filter Capsule, 1020 cm2, EFA

Catalog No. 02555453
Change view
Click to view available options
Grade:
120ZB05A
120ZB08A
120ZB10A
60ZB
60ZB05A
90ZB
90ZB08A
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
02-555-453 120ZB08A
02-555-544 120ZB05A
02-555-556 120ZB10A
02-555-561 60ZB
02-555-781 60ZB05A
02-555-456 90ZB
02-555-474 90ZB08A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
The brand will transition from 3M to Thermo Scientific
Catalog No. 02-555-453 Supplier Thermo Scientific Supplier No. E1020FSA 120ZB08A
Add to Cart
Add to Cart
The brand will transition from 3M to Thermo Scientific

Encapsulated System Scale‐Up Filter Capsules with ZB Series Media minimize hold‐up volume and maximize product recovery while delivering a particle‐free permeate solution for downstream processing.

  • Suitable for primary cell culture filtration or intermediate polishing applications in research and development, Process Development or manufacturing of recombinant protein drugs such as monoclonal antibodies
  • Made using cellulose fibers, diatomaceous earth and a binding resin that imparts a positive charge (quaternary amine), ZB grade media filters particles from process solutions through mechanical sieving and electrokinetic adsorption mechanisms
  • Wide range of filter pore sizes (0.1 to 4 μm) and a dual layer lenticular construction allows for optimizing filter throughput and filtrate quality
  • Scaleable single use filter portfolio enables using the same filtration platform process liquids volumes of 0.5 to 20,000 liters
  • Unique filter capsule design minimizes hold-up volume and maximizes product recovery, while reducing the need for cleaning validation; translucent plastic shell allows easy detection of the liquid level for real time monitoring of the filtration process and to minimize flush volumes

Specifications

Disposable Single-use
Filter Type Filter Capsule
Material (Membrane) Cellulose
Sterility Non-sterile
Grade 120ZB08A
Quantity 1 Ea.
Type Filter Capsule
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.