Learn More
Thermo Scientific Zeta Plus™ BC Series Encapsulated Disposable Filter Capsule with SP Series Media, 25 cm2, EFA, Sanitary
Description
- Zeta Plus™ SP Series is offered in the largest selection of grades in the depth filter portfolio with a range of porosity and tightness suitable for diverse applications from cell culture harvest to fine clarification
- Single layer media configurations are available in capsule and cartridge formats for high surface loading filtration applications
- BC25 Laboratory capsules contain same media as in production scale systems and are ideal for selecting the optimum media grade using low challenge fluid volume
- Full range of scalable single-use capsules and cartridges enables predictable performance from the laboratory to commercial production
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Filter Capsule |
| Material (Membrane) | Cellulose |
| Sterility | Non-sterile |
| Grade | 60SP |
| Quantity | 4/Cs. |
| Type | Filter Capsule |