Learn More
Thermo Scientific LifeASSURE™ PFS Series Filter Cartridge, 226/Spear, Silicone
Description
- Flow-enhancing construction allows air flow with a relatively low pressure drop, resulting in a smaller vent filter assemblies
- Hydrophobic polytetrafluoroethylene (PTFE) membrane helps prevent pore blockage in vent applications, extending filter lifespan
- Liquid validation of B. diminuta retention helps provide reliable sterilizing performance in wet or dry conditions
- 100% forward flow integrity tested prior to release
Specifications
Specifications
| Pore Size | 0.20 μm |
| Grade | PFS020 |
| Quantity | 6/Cs. |
| Type | Filter Cartridge |
| Length (English) | 10 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.