Learn More
Thermo Scientific Zeta Plus™ Encapsulated System Filter Capsule with LA Series Media, 170 cm2
Description
- High purity pre-extracted filter-aid and cellulose provides low extractable levels, especially aluminum
- FDA Drug Master File and USP Class VI Biological Reactivity Tests eases validation and regulatory submissions
- Self-contained, single-use capsules are available in three convenient sizes for scale-up and benchtop studies
- Low hold-up volume for minimal fluid loss
- Novel shape maintains the same lenticular flow path as larger capsules and cartridges
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Filter Capsule |
| Material (Membrane) | Cellulose |
| Sterility | Non-sterile |
| Grade | 30LA |
| Quantity | 1 Ea. |
| Type | Filter Capsule |