Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
SDP

Catalog No. 02985135 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inner:
1.3cm
3cm
Material:
Bromobutyl Rubber
Chlorobutyl Rubber
Hardness:
45 Durometer
50 Durometer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Material Hardness
02-985-135 1.3cm Chlorobutyl Rubber 45 Durometer
03-084-141 3cm Bromobutyl Rubber 50 Durometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-985-135 Supplier DWK Life Sciences Supplier No. W224100081
Add to Cart
Edge
Add to Cart

For maximum sealing of vials with blow-back glass finish

  • Made of gray bromobutyl

Not recommended for vials with a straight-walled finish.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Product Type Plug Stopper
Material Chlorobutyl Rubber
Diameter (Metric) Inner 1.3cm
For Use With (Equipment) Fits 0.7cm I.D., 1.3cm O.D. mouth of vials and serum bottles
Hardness 45 Durometer
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.