Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Screw Vial Convenience Kit

Catalog No. 03393C
Change view
Click to view available options
Volume (Metric):
4 mL
8 mL
12 mL
16 mL
Color:
Amber
Clear
Closure Type:
Screw Cap, Open Top
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Closure Type
03-393C 16 mL Clear Screw Cap, Open Top
03-393G 4 mL Clear Screw Cap, Open Top
03-393A 8 mL Clear Screw Cap, Open Top
03-393B 12 mL Clear Screw Cap, Open Top
03-391-9D 12 mL Amber Screw Cap, Open Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-393C Supplier Thermo Scientific™ Supplier No. B79904
Add to Cart
Add to Cart

Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.

Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.

  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.

Unassembled convenience kits saves time during sample preparation.

  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
TRUSTED_SUSTAINABILITY

Specifications

Description Screw Vial Convenience Kit, Open top Cap
Color Clear
Material Glass
Volume (Metric) 16 mL
Septum PTFE/Silicone
Includes 16mL clear glass vial, open top PP cap, PTFE/silicone septum
No. per Pack 200
Closure Type Screw Cap, Open Top
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.