Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling

Catalog No. 03411740 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Isotonic Saline
Machine Oil
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite (Bleach)
Toluene
Water
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Quantity:
2/Pk.
4/Pk.
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Color Quantity
03-411-740 1000 mL Ethanol Natural 2/Pk.
03-411-720 500 mL Acetone Red 4/Pk.
03-411-726 500 mL Machine Oil Natural 4/Pk.
03-411-722 500 mL Deionized Water Blue 4/Pk.
03-411-724 500 mL Ethyl Acetate Green 4/Pk.
03-411-731 500 mL Toluene Red 4/Pk.
03-411-721 500 mL Dichloromethane Yellow 4/Pk.
03-411-723 500 mL Distilled Water Blue 4/Pk.
03-411-725 500 mL Isopropanol Yellow 4/Pk.
03-411-729 500 mL Isotonic Saline Natural 4/Pk.
03-411-727 500 mL Methanol Green 4/Pk.
03-411-730 500 mL Sodium Hypochlorite (Bleach) Yellow 4/Pk.
03-411-732 500 mL Water Blue 4/Pk.
03-411-733 500 mL Ethanol Natural 4/Pk.
03-411-734 500 mL Assorted Assorted 4/Pk.
03-411-728 500 mL Methyl Ethyl Ketone Green 4/Pk.
03-411-738 1000 mL Methanol Green 2/Pk.
03-411-735 1000 mL Acetone Red 2/Pk.
03-411-737 1000 mL Isopropanol Yellow 2/Pk.
03-411-736 1000 mL Distilled Water Blue 2/Pk.
03-411-739 1000 mL Sodium Hypochlorite (Bleach) Yellow 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Catalog No. 03-411-740 Supplier Bel-Art™ Supplier No. F124320019
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position

  • Wide mouth 53 mm (2 in.) for easy refilling
  • Translucent LDPE bottles with color-coded caps for easy identification
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve
  • Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Natural
Material LDPE
Quantity 2/Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Ethanol
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.