Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Ethylphenol

Catalog No. 04688100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-688-100ML 100 mL
04-688-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-688-100ML Supplier Sigma Aldrich Supplier No. 04688100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Ethylphenol

Specifications

CAS 620-17-7
Density 1.001 g/mL (at 25°C (literature))
Molecular Formula C8H10O
Refractive Index n20/D 1.533 (literature); n20/D 1.534
Linear Formula C2H5C6H4OH
MDL Number MFCD00002311
Quantity 100 mL
Molecular Weight (g/mol) 122.16
Percent Purity ≥95.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.