Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders
GSA_VA

Catalog No. 087692B Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Size (Metric):
16 mm Spreader Bar
25 mm Spreader Bar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
08-769-2B 25 mm Spreader Bar
08-769-2A 16 mm Spreader Bar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-769-2B Supplier Bel-Art™ Supplier No. F377360009
Add to Cart
Add to Cart

Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.

  • Autoclavable and may be flame sterilized
  • Features triangular end with smooth rounded surfaces throughout
  • Smaller 16mm width bar suitable for use with 6cm plates
  • Larger 25mm width bar suitable for use with 9cm plates
  • Use with Inoculating Turntable (Mfr. No.F40913-0000)
  • Both sizes 30cm long

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Size (Metric) 25 mm Spreader Bar
Autoclavable Autoclavable
Length (Metric) 30.2 cm
Sterility Sterile
Length (English) 12 in.
Product Type Bacterial Cell Spreader
For Use With (Equipment) Inoculating turntable, 9cm inoculating plates
Material Stainless Steel
For Use With (Application) Easily perform inoculation of petri dishes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.