Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ MD 100 Chlorine Colorimeter

Catalog No. 09053542 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Chlorine Colorimeter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-053-542 Chlorine Colorimeter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-542 Supplier Lovibond™ Supplier No. 276010
Add to Cart
Add to Cart

The MD 100 Chlorine Colorimeter is a single parameter instrument for measuring the concentration of Free and Total Chlorine in a water sample. The hand-held design makes it ideal for use outside of a laboratory setting and the simplistic user interface ensures ease-of-use by operators of all levels.

  • Waterproof Design: The IP 68 design of this instrument ensures that the MD 100 stays watertight for up to 1 hour at 0.1 meter. Don't worry about dropping the instrument into a body of water - it floats.
  • Accurate, Reproducible Results: The high-quality optical system of the MD 100 quickly delivers results that you can be confident in. Secondary standards are available for on-site verification of instrument performance.
  • User Calibration Mode: A one-point calibration is possible, and all MD 100 instruments can display readings directly in ABS or %T units.
  • One-time-Zero Function: Don't waste time waiting for your instrument to zero after each test. When testing a new sample, zero the instrument once and all subsequent tests of that sample do not require you to rezero the instrument.
  • Automatic Data Storage: The MD 100 automatically stores the last 16 readings with time and date stamp. These results can be transferred to a spreadsheet using the IRiM accessory.
  • Ergonomic Design: Whether you're using the MD 100 out in the field or in the lab, the design of the instrument makes it easy to handle and view the display. If you're using the instrument in the lab, the rubber feet on the bottom of the housing eliminate the need for two hand operation.
  • Factory Programmed: All MD 100 instruments are supplied ready to use with pre-programmed calibration curves. All you need to do is select the measuring range you need, prepare your sample and test.
  • Scroll Driven Menu System: The simple interface allows you to quickly reach the test you need. When the instrument is turned on, it will automatically display the last test used.

Specifications

Accuracy 3% Full scale
Backlight Option Yes
Battery Life 17 Hours or ∼5000 tests
Calibration Factory Programmed & Calibrated
Certifications/Compliance CE
Cuvette Details 24 mm, glass for low range method; 10 mm, plastic cells for high range method
Data Storage Automatic; 16 data sets with time and date stamp
Battery Type AA
Display LCD
Dimensions (L x W x H) 15.5 x 7.6 x 3.8 cm (6.1 x 3 x 1.5 in.)
IP Rating IP68
Languages Instrument: English, Manual: English, French, Spanish, Portuguese
Light Source LED
Type Chlorine Colorimeter
Wavelength Selection Automatic
For Use With (Application) Measuring chlorine residual outside of a laboratory setting
Height (English) 1.5 in.
Height (Metric) 3.8 cm
Includes Instrument, 3 x 24 mm sample cells, 2 x Type 2 multi-vials, DPD free chlorine reagent, 10 mL sample (100 Pk.), DPD total chlorine reagent, 10 mL sample (100 Pk.), batteries, tube brush, instruction manual and a carrying case
Length (English) 6.1 in.
Length (Metric) 15.5 cm
Material ABS
Measurement Range 0 to 2.0 mg/L, 0 to 8.0 mg/L Cl2
Model MD 100
Outputs Yes, with additional accessory
Packaging Powder Pack
Range 0 to 2.0 mg/L, 0 to 8.0 mg/L
Resolution 0.0001
Sample Size 10 mL
Temperature Range (Metric) 5°C to 40°C
Test Method DPD
Warranty 2 Years
Wavelength 530 nm
Wavelength Accuracy ± 5 nm
Weight (English) 0.55 lb.
Weight (Metric) 250 g
Width (English) 3 in.
Width (Metric) 7.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.