Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Blue Starline Polypropylene Caps for Narrow-Mouth Containers
SDP

Catalog No. 0987713D Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
20-410
24-410
28-410
38-430
No. per Case:
48 Cs.
72 Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
09-877-13D 28-410 48 Cs.
09-877-11D 20-410 72 Cs.
09-877-14D 38-430 72 Cs.
09-877-12D 24-410 72 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-877-13D Supplier DWK Life Sciences Supplier No. 23951204
Add to Cart
Add to Cart

Replacement caps for Leak-Resistant Containers

  • Leak-proof design
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
No. per Case 48 Cs.
Neck Finish 28-410
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.