Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone 98.0+%, TCI America™
SDP

Catalog No. N1038100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1038100G 100 g
N103825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1038100G Supplier TCI America Supplier No. N1038100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 756-13-8
Molecular Formula C6F12O
Molecular Weight (g/mol) 316.05
MDL Number MFCD04038343
InChI Key RMLFHPWPTXWZNJ-UHFFFAOYSA-N
Synonym Perfluoroethyl Perfluoroisopropyl Ketone
PubChem CID 2782408
IUPAC Name 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
SMILES FC(F)(F)C(F)(F)C(=O)C(F)(C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Quantity 100 g
Formula Weight 316.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone
EINECSNumber (2)-4024
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.