Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%

Catalog No. AAA1274714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274714 25 g
AAA1274706 5 g
AAA1274722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1274714 Supplier Thermo Scientific Chemicals Supplier No. A1274714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 920-66-1 | C3H2F6O | 168.038 g/mol

1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

Solubility
Miscible with water, chloroform, ether and acetone.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases, aluminum, zinc and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point -4°C
Density 1.619
Boiling Point 59°C to 60°C
Refractive Index 1.275
Linear Formula CF3CH(OH)CF3
Quantity 25 g
UN Number UN1760
Beilstein 1841007
Solubility Information Miscible with water,chloroform,ether and acetone.
Formula Weight 168.04
Percent Purity ≥99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 213-059-4
RTECSNumber UB6450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.