accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFF

CAS RN 920-66-1

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-ol
Synonyms: hexafluoroisopropanol hexafluoroisopropyl alcohol 2H-hexafluoroisopropanol HFIP hexafluoro-2-propanol
Molecular Weight (g/mol): 168.04
Molecular Formula: C3H2F6O
InChi Key: BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES: OC(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (18)
  • (14)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (5)
  • (5)
  • (7)
  • (2)
  • (3)
  • (11)
Percent Purity 
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (10)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (20)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (5)
  • (1)
  • (2)
  • (2)
  • (5)
  • (5)
  • (1)
  • (3)
  • (18)
  • (2)
  • (18)
  • (14)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (5)
  • (7)
  • (2)
  • (3)
  • (11)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (10)
  • (1)
  • (1)
  • (2)
  • (20)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (5)
  • (4)
  • (1)