Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)

Catalog No. AA4229103
Change view
Click to view available options
Quantity:
1 g
5 x 1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4229103 1 g
AA4229146 5 x 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4229103 Supplier Thermo Scientific Chemicals Supplier No. 04229103
Add to Cart

CAS: 38701-74-5 | C3H2F6O | 170.05 g/mol

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2} is a stable isotope used for biochemical research. It is also used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38701-74-5
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 170.05
MDL Number MFCD00037559
InChI Key BYEAHWXPCBROCE-AWPANEGFSA-N
PubChem CID 2724852
SMILES [2H]OC([2H])(C(F)(F)F)C(F)(F)F

Specifications

Physical Form Liquid
Quantity 1 g
Packaging Prescored Ampule
Sensitivity Moisture
Density 1.60 g/mL
Melting Point −4°C
Boiling Point 59°C
UN Number UN1760
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 1,1,1,3,3,3-hexafluoro 2-2h propan-2-2h ol, hexafluoro 2 h propan-2-2 h ol, hexafluoro 2h propan-2-2h ol, 1,1,1,3,3,3-hexafluoro-2-propanol-d2 hfip-d2, 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 99 atom % d
Solubility Information Fully miscible in water.
IUPAC Name 2-deuterio-2-deuteriooxy-1,1,1,3,3,3-hexafluoropropane
Formula Weight 170.05
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2, Isotopic
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 254-091-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.