Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, LC-MS Grade, Solstice

Catalog No. 60046851 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-851 10 x 1 mL Ampoule
60-046-852 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-851 Supplier Solstice Supplier No. 4206010X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point -4°C
Color Colorless
Density 1.596g/mL at 25°C
Boiling Point 59°C
Packaging Ampoule
Refractive Index n20/D 1.275
Linear Formula (CF3)2CHOH
Quantity 10 x 1 mL
UN Number UN3265
Beilstein 1841007
Residue after Evaporation 0 to 0.0005%
Formula Weight 168.04g/mol
Percent Purity ≥99.8% (GC)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8/PGII
RTECSNumber 213-059-4
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.