Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600018 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-018 5 kg Amber Glass Bottle
18-600-014 5 g Amber Glass Bottle
18-600-015 25 g Amber Glass Bottle
18-600-016 100 g Amber Glass Bottle
18-600-017 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-018 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H10085KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

H1008, 920-66-1, C3H2F6O

Spectrum™ Chemical 1,1,1,3,3,3-Hexafluoro-2-propanol, Reagent , is the organic compound that is commonly abbreviated as HFIP and is a fluorinated alcohol that is used as a solvent and synthetic intermediate. It′s a colorless, volatile liquid that has a characteristic strong, pungent odor. It is mainly used for solubilizing a rangeof polymers as well as being the precursor and chief metabolite of the inhalation anesthetic sevoflurane. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.04
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES OC(C(F)(F)F)C(F)(F)F

Specifications

CAS Max % 100%
Boiling Point 57°C to 60°C/134.6°F to 140°F
Packaging Amber Glass Bottle
Quantity 5 kg
UN Number UN1760
Percent Purity 99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.