Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoroacetone sesquihydrate, 98%

Catalog No. ac269970250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269970250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC269970250 Supplier Thermo Scientific Chemicals Supplier No. 269970250
Only null left
Add to Cart

CAS: 13098-39-0 | 1·5 H2O | 193.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13098-39-0
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 193.05
MDL Number MFCD06800792
InChI Key VAIZVCMDJPBJCM-UHFFFAOYSA-N
Synonym hexafluoroacetone sesquihydrate, unii-cu1noz8ps6, ccris 4653, acetone, hexafluoro-, sesquihydrate, hexafluoro-2-propanone sesquihydrate, cu1noz8ps6, 2-propanone, hexafluoro-, sesquihydrate, 1,1,1,3,3,3-hexafluoroacetone sesquihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, hydrate 2:3, hexafluoroacetone 1.5-hydrate
PubChem CID 3032602
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES C(=O)(C(F)(F)F)C(F)(F)F.C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C to 21°C
Color Amber or Yellow
Density 1.6900g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Plastic bottle
Refractive Index 1.3060 to 1.311
Quantity 25 g
Specific Gravity 1.69
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
Viscosity 0.67 mPa.s (38°C)
Formula Weight 193.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone sesquihydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.