Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane 96.0+%, TCI America™
SDP

Catalog No. H126625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H126625G 25 g
H12665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H126625G Supplier TCI America Supplier No. H126625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7422-52-8
Molecular Formula C13H32O4Si3
Molecular Weight (g/mol) 336.65
MDL Number MFCD00053926
InChI Key YSIQPJVFCSCUMU-UHFFFAOYSA-N
PubChem CID 62635
IUPAC Name trimethyl-[methyl-[3-(oxiran-2-ylmethoxy)propyl]-trimethylsilyloxysilyl]oxysilane
SMILES C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 g
Formula Weight 336.65
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.