Learn More
Sigma Organic Chemistry 1 1 1-TrifluOR-2 4-hexa | 500mg | 400-54-4 | MFCD00051306
Supplier: Sigma Organic Chemistry CDS003384500MG

1 1 1-TrifluOR-2 4-hexa , 500mg
About this Item:
CAS #: 400-54-4
MDL #: MFCD00051306
Molecular Weight: 168.11
UNSPSC Code: 12352200
Chemical Formula: C6H7F3O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.