Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Tris(4-hydroxyphenyl)ethane 98.0+%, TCI America™
SDP

Catalog No. T1254100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1254100G 100 g
T125425G 25 g
T1254500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1254100G Supplier TCI America Supplier No. T1254100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27955-94-8
Molecular Formula C20H18O3
Molecular Weight (g/mol) 306.361
MDL Number MFCD00012180
InChI Key BRPSWMCDEYMRPE-UHFFFAOYSA-N
PubChem CID 93118
IUPAC Name 4-[1,1-bis(4-hydroxyphenyl)ethyl]phenol
SMILES CC(C1=CC=C(C=C1)O)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O

Specifications

Melting Point 248°C
Color White
Quantity 100 g
UN Number 3077
Formula Weight 306.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,1-Tris(4-hydroxyphenyl)ethane
EINECSNumber (4)-1668
RTECSNumber SL4375275
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.