Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600021 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-021 1 L Amber Glass Bottle
18-600-019 250 mL Amber Glass Bottle
18-600-022 4 L Amber Glass Bottle
18-600-020 20 L Steel Can
18-600-023 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-021 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T10181LTGL
Only null left
Add to Cart
Add to Cart

T1018, 79-34-5, C2H2Cl4

Spectrum™ Chemical 1,1,2,2-Tetrachloroethane , also known as Acetylene tetrachloride, is a chlorinated derivative of ethane with a very high solvent power. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.84
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES ClC(Cl)C(Cl)Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 L
UN Number UN1702
Percent Purity 98%
Grade Reagent
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.