Learn More
1,1,2,2-Tetrachloroethane, 98+%
Description
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.
Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
Specifications
Specifications
| Melting Point | -43°C |
| Density | 1.596 |
| Boiling Point | 142°C to 146°C |
| Refractive Index | 1.4935 |
| Linear Formula | Cl2CHCHCl2 |
| Quantity | 100 g |
| UN Number | UN1702 |
| Beilstein | 969206 |
| Merck Index | 14,9189 |
| Solubility Information | Miscible with water,methanol,ethanol,chloroform and acetone. |
| Show More |
Safety and Handling
| Hazard Category | H310+H330-H351 |
| Hazard Statement | GHS H Statement H310-H330-H341-H351 Fatal in contact with skin. Fatal if inhaled. Suspected of causing genetic defects. Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P260-P262-P264b-P270-P271-P281-P284-P302+P350-P304+P340-P308+P313-P310-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE |
| EINECSNumber | 201-197-8 |
| RTECSNumber | KI8575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.