Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,3,3,3-Hexafluoropropyl Methyl Ether 98.0+%, TCI America™
SDP

Catalog No. H150725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H150725G 25 g
H15075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H150725G Supplier TCI America Supplier No. H150725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 382-34-3
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00041537
InChI Key PKMXTDVNDDDCSY-UHFFFAOYSA-N
PubChem CID 2778364
IUPAC Name 1,1,1,2,3,3-hexafluoro-3-methoxypropane
SMILES COC(C(C(F)(F)F)F)(F)F

Specifications

Color Colorless
Boiling Point 55°C
Quantity 25 g
UN Number 3271
Formula Weight 182.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,3,3,3-Hexafluoropropyl Methyl Ether
EINECSNumber (2)-3998
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.