Learn More
Sigma Aldrich 1,1,2-Trichloroethane
Description
- 1,1,2-trichloroethane (1,1,2-tca), a chlorinated aliphatic hydrocarbon, is a common groundwater pollutant
- Microbial detoxification of 1,1,2-tca has been studied
- Ni supported on mesoporous silica (sba-16) catalyzed hydrodechlorination of 1,1,2-trichloroethane has been reported
- Rotational isomers of 1,1,2-trichloroethane in the gaseous and liquid states have been investigated by the infrared absorption spectra
Specifications
Specifications
| CAS | 79-00-5 |
| Density | 1.435 g/mL (at 25°C (literature)) |
| Boiling Point | 110°C to 115°C (lit.) |
| Molecular Formula | C2H3Cl3 |
| Refractive Index | n20/D 1.471 (literature) |
| Linear Formula | ClCH2CHCl2 |
| MDL Number | MFCD00000852 |
| Quantity | 25 mL |
| Molecular Weight (g/mol) | 133.4 |
| Percent Purity | 97% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.