Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane 95.0+%, TCI America™
SDP

Catalog No. T35481ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35481ML 1 mL
T35485ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35481ML Supplier TCI America Supplier No. T35481ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19095-23-9
Molecular Formula C14H44O6Si7

Specifications

Color Colorless
Boiling Point 128°C
Quantity 1 mL
Formula Weight 505.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.