Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane 97.0+%, TCI America™
SDP

Catalog No. H144125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H144125ML 25 mL
H14415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H144125ML Supplier TCI America Supplier No. H144125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1189-93-1
Molecular Formula C6H18O2Si3
Molecular Weight (g/mol) 206.463
MDL Number MFCD00039788
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
PubChem CID 6327152
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C

Specifications

Boiling Point 128°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
UN Number 1993
Formula Weight 208.48
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
EINECSNumber (7)-0477
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.