Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3-Tetramethoxypropane, 98%

Catalog No. AAA114180B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA114180B 1000 g
AAA1141830 250 g
AAA114180I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA114180B Supplier Thermo Scientific Chemicals Supplier No. A114180B
Only null left
Add to Cart

CAS: 102-52-3 | C7H16O4 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-52-3
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008488
InChI Key XHTYQFMRBQUCPX-UHFFFAOYSA-N
Synonym propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
PubChem CID 66019
IUPAC Name 1,1,3,3-tetramethoxypropane
SMILES COC(CC(OC)OC)OC

Specifications

Density 0.997
Boiling Point 182°C to 183°C
Flash Point 54°C (129°F)
Refractive Index 1.407
Quantity 1000 g
UN Number UN3271
Beilstein 1700020
Sensitivity Moisture sensitive
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 1,1,3,3-Tetramethoxypropane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 203-037-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.