Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,5-Tetramethylcyclohexane 96.0+%, TCI America™
SDP

Catalog No. T17175ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17175ML Supplier TCI America Supplier No. T17175ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4306-65-4
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00060820
InChI Key WOJSMJIXPQLESQ-UHFFFAOYSA-N
PubChem CID 107263
IUPAC Name 1,1,3,5-tetramethylcyclohexane
SMILES CC1CC(CC(C1)(C)C)C

Specifications

Color Colorless
Boiling Point 148°C
Quantity 5 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,5-Tetramethylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.