Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-(Azodicarbonyl)dipiperidine 98.0+%, TCI America™
SDP

Catalog No. A105125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A105125G 25 g
A10515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A105125G Supplier TCI America Supplier No. A105125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10465-81-3
Molecular Formula C12H20N4O2
Molecular Weight (g/mol) 252.32
MDL Number MFCD00010111
InChI Key OQJBFFCUFALWQL-UHFFFAOYSA-N
Synonym 1,1'-azodicarbonyl-dipiperidine, 1,1-azodicarbonyl dipiperidine, 1,1'-azodicarbonyl dipiperidine, addp, diazene-1,2-diylbis piperidin-1-ylmethanone, azodicarboxylic acid dipiperidine, ne-n-piperidine-1-carbonylimino piperidine-1-carboxamide, 1,1'-azodicarbonyl dipiperidine addp, piperidyl piperidylcarbonyl diazenyl ketone, e-diazene-1,2-diylbis piperidin-1-ylmethanone
PubChem CID 5702657
IUPAC Name N-[(piperidine-1-carbonyl)imino]piperidine-1-carboxamide
SMILES O=C(N=NC(=O)N1CCCCC1)N1CCCCC1

Specifications

Melting Point 136°C
Quantity 25 g
Formula Weight 252.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(Azodicarbonyl)dipiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.