Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Benzofuran-2-yl)-2-bromoethan-1-one, 90%, Thermo Scientific™

Catalog No. CC06605CB
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC06605CB 250 mg
CC06605DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC06605CB Supplier Thermo Scientific Chemicals Supplier No. CC06605CB

Chemical Identifiers

CAS 23489-36-3
Molecular Formula C10H7BrO2
Molecular Weight (g/mol) 239.068
InChI Key NVRNCBWTEDOAQA-UHFFFAOYSA-N
Synonym 1-1-benzofuran-2-yl-2-bromoethan-1-one, 1-1-benzofuran-2-yl-2-bromoethanone, 2-2-bromoacetyl benzofuran, 1-benzofuran-2-yl-2-bromoethanone, 1-benzofuran-2-yl-2-bromo-ethanone, 2-bromoacetylbenzofuran, 2-bromoacetyl benzofuran, 1-benzo d furan-2-yl-2-bromoethan-1-one, pubchem7013, 2-bromoacetylbenzo b furan
PubChem CID 2735451
IUPAC Name 1-(1-benzofuran-2-yl)-2-bromoethanone
SMILES C1=CC=C2C(=C1)C=C(O2)C(=O)CBr

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 239.07
Percent Purity 90%
Chemical Name or Material 1-(1-Benzofuran-2-yl)-2-bromoethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.