Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene 98.0+%, TCI America™
SDP

Catalog No. B35011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35011G 1 g
B35015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35011G Supplier TCI America Supplier No. B35011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 737776-93-1
Molecular Formula C22H32B2FeO4
MDL Number MFCD06411336

Specifications

Melting Point 173°C
Quantity 1 g
Synonym 1,1′C-Ferrocenediboronic Acid Bis(pinacol) Ester
Formula Weight 437.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.