Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) Dichloride 97.0+%, TCI America™
SDP

Catalog No. B22261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22261G 1 g
B22265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22261G Supplier TCI America Supplier No. B22261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67292-34-6
Molecular Formula C34H28Cl2FeNiP2
Molecular Weight (g/mol) 683.98
MDL Number MFCD00270284
IUPAC Name Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
SMILES [Fe].Cl[Ni]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 259°C
Quantity 1 g
Synonym [1,1′C-Bis(diphenylphosphino)ferrocene]dichloronickel(II), Dichloro[1,1′C-bis(diphenylphosphino)ferrocene]nickel(II)
Formula Weight 683.98
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,1′-Bis(diphenylphosphino)ferrocene]nickel(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.