Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diphenylphosphino)ferrocene 96.0+%, TCI America™
SDP

Catalog No. B20271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20271G 1 g
B20275G 5 g
B202725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B20271G Supplier TCI America Supplier No. B20271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12150-46-8
Molecular Formula C34H28FeP2
Molecular Weight (g/mol) 554.39
MDL Number MFCD00001422
InChI Key KZPYGQFFRCFCPP-UHFFFAOYSA-N
IUPAC Name λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
SMILES [Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 182°C
Quantity 1 g
Synonym 1,1′C-Ferrocenebis(diphenylphosphine)
Formula Weight 554.39
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(diphenylphosphino)ferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.